Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lancl3
Entrez Gene ID
236285
Gene Name
LanC lantibiotic synthetase component C-like 3 (bacterial)
Gene Aliases
6030463G20Rik
Chromosome Location
Chr.X: 9199973 - 9268085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173414.3 NP_775590.2 2 - 3 951 123
GenBank mRNA AK031623.1 - 2 - 3 952 123
AK043841.1 - 2 - 3 152 123
AK045254.1 - 2 - 3 124

View Full Product Details