Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp7b1
Entrez Gene ID
13123
Gene Name
cytochrome P450, family 7, subfamily b, polypeptide 1
Gene Aliases
AW261589, D3Ertd552e, hct-1
Chromosome Location
Chr.3: 18071950 - 18243893 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007825.4 NP_031851.3 5 - 6 1360 151
GenBank mRNA AK013034.1 - 5 - 6 1290 151
AK140100.1 - 5 - 6 1352 151
AK172152.1 - 5 - 6 1361 151
BC038810.1 - 5 - 6 1320 151
U36993.1 - 5 - 6 1305 151

View Full Product Details