The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081304.1 | NP_001074773.1 | 13 - 14 | 1634 | 74 |
GenBank mRNA | AK020270.1 | - | 13 - 14 | 1615 | 74 |
AK036106.1 | - | 13 - 14 | 1613 | 74 | |
AK036335.1 | - | 13 - 14 | 1632 | 74 | |
AK087000.1 | - | 13 - 14 | 1620 | 74 | |
BC043662.1 | - | 12 - 13 | 1481 | 74 | |
BC141028.1 | - | 13 - 14 | 1628 | 74 |