Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atf6
Entrez Gene ID
226641
Gene Name
activating transcription factor 6
Gene Aliases
9130025P16Rik, 9630036G24, AA789574, Atf6alpha, ESTM49
Chromosome Location
Chr.1: 170704457 - 170867771 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081304.1 NP_001074773.1 2 - 3 196 92
GenBank mRNA AK020270.1 - 2 - 3 176 92
AK036106.1 - 2 - 3 176 92
AK036335.1 - 2 - 3 194 92
AK087000.1 - 2 - 3 177 92
BC043662.1 - 1 - 2 43 92
BC141028.1 - 2 - 3 190 92

View Full Product Details