Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcaf6
Entrez Gene ID
74106
Gene Name
DDB1 and CUL4 associated factor 6
Gene Aliases
1200006M05Rik, Iqwd1, PC326
Chromosome Location
Chr.1: 165329500 - 165460553 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028759.1 NP_083035.1 11 - 12 1731 127
GenBank mRNA AK004618.1 - 11 - 12 1594 127
AK077238.1 - 2 - 3 249 127
BC058980.1 - 9 - 10 1294 127
BC084732.1 - 9 - 10 1294 127

View Full Product Details