Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gm9
Entrez Gene ID
194854
Gene Name
predicted gene 9
Gene Aliases
-
Chromosome Location
Chr.X: 37208502 - 37211041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033234.2 NP_001028406.1 2 - 3 535 69
GenBank mRNA AK131663.1 - 2 - 3 500 69
AK145837.1 - 2 - 3 536 69
BC048624.1 - 2 - 3 499 69

View Full Product Details