Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rsad1
- Entrez Gene ID
-
237926
- Gene Name
- radical S-adenosyl methionine domain containing 1
- Gene Aliases
-
B430319G23, BC056485
- Chromosome Location
-
Chr.11: 94539798 - 94556861 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AW060343.1 |
- |
1 - 2 |
|
92 |
View Full Product Details