The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007405.2 | NP_031431.2 | 8 - 9 | 1833 | 88 |
GenBank mRNA | AK147516.1 | - | 8 - 9 | 1833 | 88 |
BC145101.1 | - | 7 - 8 | 1666 | 88 | |
M93422.1 | - | 7 - 8 | 1765 | 88 | |
M96653.1 | - | 7 - 8 | 1655 | 88 |