The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007622.3 | NP_031648.1 | 4 - 5 | 667 | 82 |
GenBank mRNA | AK014215.1 | - | 4 - 5 | 643 | 82 |
X56690.1 | - | 3 - 4 | 412 | 82 |