Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Card6
Entrez Gene ID
239319
Gene Name
caspase recruitment domain family, member 6
Gene Aliases
D730008L15
Chromosome Location
Chr.15: 5096920 - 5116483 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163138.1 NP_001156610.1 3 - 4 873 121

View Full Product Details