Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cntnap3
Entrez Gene ID
238680
Gene Name
contactin associated protein-like 3
Gene Aliases
-
Chromosome Location
Chr.13: 64736806 - 64904028 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081129.1 NP_001074598.1 6 - 7 982 121

View Full Product Details