The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081062.1 | NP_001074531.1 | 1 - 2 | 483 | 80 |
GenBank mRNA | AK086507.1 | - | 1 - 2 | 458 | 80 |
BC147759.1 | - | 1 - 2 | 459 | 80 | |
BC147766.1 | - | 1 - 2 | 459 | 80 |