Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ano6
Entrez Gene ID
105722
Gene Name
anoctamin 6
Gene Aliases
2900059G15Rik, AA407480, AW554778, F730003B03Rik, Tmem16f
Chromosome Location
Chr.15: 95790843 - 95975472 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253813.1 NP_001240742.1 18 - 19 2430 118
NM_175344.4 NP_780553.2 17 - 18 2367 118
GenBank mRNA AK089300.1 - 18 - 19 2503 118
AK159411.1 - 18 - 19 118
BC060732.1 - 17 - 18 2367 118

View Full Product Details