Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmed8
Entrez Gene ID
382620
Gene Name
transmembrane emp24 domain containing 8
Gene Aliases
6430595O10Rik, AI447224, Gm1184, Mem1
Chromosome Location
Chr.12: 87166242 - 87200229 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033475.3 NP_001028647.1 5 - 6 795 121
GenBank mRNA AK147378.1 - 5 - 6 796 121
BC150860.1 - 5 - 6 866 121

View Full Product Details