The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029934.1 | NP_001025105.1 | 33 - 34 | 4830 | 73 |
GenBank mRNA | AK032362.1 | - | 2 - 3 | 127 | 73 |
AK140739.1 | - | 2 - 3 | 203 | 73 | |
BC040268.1 | - | 10 - 11 | 1795 | 73 | |
BC051399.1 | - | 10 - 11 | 1868 | 73 | |
BC058994.1 | - | 14 - 15 | 2371 | 73 |