The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177564.5 | NP_808232.2 | 1 - 2 | 249 | 78 |
GenBank mRNA | AK157041.1 | - | 1 - 2 | 234 | 78 |
BC022224.1 | - | 1 - 2 | 210 | 78 |