Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fgd2
- Entrez Gene ID
-
26382
- Gene Name
- FYVE, RhoGEF and PH domain containing 2
- Gene Aliases
-
Tcd-2, Tcd2, ZFYVE4, tcs-2, tcs2
- Chromosome Location
-
Chr.17: 29360908 - 29379535 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
DQ344523.1 |
- |
13 - 14 |
1463 |
58 |
View Full Product Details