Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gip
Entrez Gene ID
14607
Gene Name
gastric inhibitory polypeptide
Gene Aliases
-
Chromosome Location
Chr.11: 96024534 - 96031296 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008119.2 NP_032145.2 1 - 2 76 69
GenBank mRNA AK008308.1 - 1 - 2 76 69

View Full Product Details