The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008119.2 | NP_032145.2 | 5 - 6 | 527 | 65 |
GenBank mRNA | AK008308.1 | - | 5 - 6 | 527 | 65 |
AK008525.1 | - | 4 - 5 | 383 | 65 | |
BC104314.1 | - | 5 - 6 | 517 | 65 | |
BC104315.1 | - | 5 - 6 | 517 | 65 |