Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pigs
Entrez Gene ID
276846
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class S
Gene Aliases
BC058979, Gm393, Gm689
Chromosome Location
Chr.11: 78328422 - 78342781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201406.1 NP_958808.1 3 - 4 339 71
GenBank mRNA AK033331.1 - 3 - 4 313 71
AK039604.1 - 3 - 4 310 71
AK043232.1 - 3 - 4 313 71
AK045759.1 - 3 - 4 310 71
AK053771.1 - 3 - 4 313 71
AK142871.1 - 3 - 4 310 71
AK151858.1 - 3 - 4 301 71
AK152229.1 - 3 - 4 301 71
AK153300.1 - 3 - 4 305 71
AK158128.1 - 3 - 4 310 71
AK166408.1 - 3 - 4 316 71
AK169633.1 - 3 - 4 305 71
BC058979.1 - 3 - 4 339 71

View Full Product Details