Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mllt6
Entrez Gene ID
246198
Gene Name
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6
Gene Aliases
AI315037, Af17
Chromosome Location
Chr.11: 97663412 - 97685461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139311.2 NP_647472.2 8 - 9 1020 99
GenBank mRNA AK143477.1 - 4 - 5 463 99
BY731745.1 - 4 - 5 459 99

View Full Product Details