Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc23l
Entrez Gene ID
75777
Gene Name
tetratricopeptide repeat domain 23-like
Gene Aliases
4930401A09Rik
Chromosome Location
Chr.15: 10499155 - 10559261 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029430.1 NP_083706.1 3 - 4 382 84
GenBank mRNA AK015027.1 - 3 - 4 354 84
BC132384.1 - 3 - 4 382 84
BC145852.1 - 3 - 4 382 84

View Full Product Details