Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nln
Entrez Gene ID
75805
Gene Name
neurolysin (metallopeptidase M3 family)
Gene Aliases
4930472G13Rik, C79345
Chromosome Location
Chr.13: 104023050 - 104109660 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029447.2 NP_083723.1 4 - 5 719 79
GenBank mRNA AK079090.1 - 4 - 5 720 79
AK138399.1 - 4 - 5 692 79
AK146364.1 - 4 - 5 668 79
AK170508.1 - 4 - 5 720 79
AK171432.1 - 4 - 5 720 79
BC016224.1 - 4 - 5 692 79

View Full Product Details