The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146027.2 | NP_666139.1 | 7 - 8 | 1227 | 122 |
GenBank mRNA | AK075869.1 | - | 7 - 8 | 1191 | 122 |
BC010784.1 | - | 6 - 7 | 1271 | 122 | |
BC021346.1 | - | 7 - 8 | 1196 | 122 |