The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205345.1 | NP_001192274.1 | 2 - 3 | 363 | 66 |
NM_001205346.1 | NP_001192275.1 | 2 - 3 | 363 | 66 | |
NM_194342.3 | NP_919323.2 | 2 - 3 | 363 | 66 | |
GenBank mRNA | AK046318.1 | - | 2 - 3 | 332 | 66 |
AK085196.1 | - | 2 - 3 | 332 | 66 | |
AK128958.1 | - | 2 - 3 | 357 | 66 | |
AK156246.1 | - | 2 - 3 | 364 | 66 | |
AK171058.1 | - | 2 - 3 | 332 | 66 | |
AK172398.1 | - | 2 - 3 | 364 | 66 | |
AK172996.1 | - | 2 - 3 | 188 | 66 | |
AY682987.1 | - | 1 - 2 | 129 | 66 | |
BC098208.1 | - | 2 - 3 | 283 | 66 |