The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207225.1 | NP_997108.1 | 24 - 25 | 3167 | 110 |
GenBank mRNA | AK029933.1 | - | 21 - 22 | 2719 | 110 |
AK155250.1 | - | 25 - 26 | 3353 | 110 | |
AK162369.1 | - | 2 - 3 | 225 | 110 | |
BC066052.1 | - | 24 - 25 | 3167 | 110 |