The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134252.3 | NP_599013.1 | 11 - 12 | 1422 | 61 |
GenBank mRNA | AF481480.2 | - | - | 1691 | 61 |
AY095352.1 | - | 9 - 10 | 1256 | 61 | |
BC117934.1 | - | 11 - 12 | 1422 | 61 |