The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162868.1 | NP_001156340.1 | 1 - 2 | 1594 | 60 |
NM_001162869.1 | NP_001156341.1 | 1 - 2 | 1594 | 60 | |
GenBank mRNA | AB093257.1 | - | 1 - 2 | 1035 | 60 |
AK080103.1 | - | 2 - 3 | 1709 | 60 | |
AK083658.1 | - | 1 - 2 | 1278 | 60 |