The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024716.1 | NP_001019887.1 | 2 - 3 | 204 | 78 |
NM_001039156.1 | NP_001034245.1 | 13 - 14 | 4971 | 78 | |
NM_138579.4 | NP_613045.3 | 12 - 13 | 4833 | 78 | |
GenBank mRNA | AK053194.1 | - | 2 - 3 | 164 | 78 |
AK085512.1 | - | 2 - 3 | 174 | 78 | |
AK129417.1 | - | 2 - 3 | 154 | 78 | |
AK170390.1 | - | 2 - 3 | 150 | 78 | |
BC042760.1 | - | 2 - 3 | 143 | 78 | |
BC053329.1 | - | 2 - 3 | 188 | 78 | |
CF533172.1 | - | 2 - 3 | 276 | 78 | |
DQ228000.1 | - | 13 - 14 | 4971 | 78 | |
DQ228001.2 | - | 12 - 13 | 4833 | 78 |