The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040187.1 | NP_001035277.1 | 1 - 2 | 553 | 105 |
NM_199079.1 | NP_951061.1 | 1 - 2 | 372 | 105 | |
NM_199080.2 | NP_951062.1 | 1 - 2 | 553 | 105 | |
GenBank mRNA | AK039888.1 | - | 1 - 2 | 372 | 105 |
AK151061.1 | - | 1 - 2 | 328 | 105 | |
AK152749.1 | - | 1 - 2 | 328 | 105 | |
AK152831.1 | - | 1 - 2 | 328 | 105 | |
AK161001.1 | - | 1 - 2 | 369 | 105 | |
AK161012.1 | - | 1 - 2 | 368 | 105 | |
BC062910.1 | - | 1 - 2 | 358 | 105 | |
BC096036.1 | - | 1 - 2 | 553 | 105 |