The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081315.1 | NP_001074784.1 | 11 - 12 | 3538 | 138 |
GenBank mRNA | AK129322.1 | - | 9 - 10 | 1793 | 138 |
AK147555.1 | - | 11 - 12 | 3537 | 138 | |
AK162638.1 | - | 4 - 5 | 386 | 138 | |
BC157915.1 | - | 10 - 11 | 3303 | 138 |