Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brpf3
Entrez Gene ID
268936
Gene Name
bromodomain and PHD finger containing, 3
Gene Aliases
AI413466, AW493400, mKIAA1286
Chromosome Location
Chr.17: 28800557 - 28838789 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081315.1 NP_001074784.1 4 - 5 1989 61
GenBank mRNA AK129322.1 - 2 - 3 244 61
AK147555.1 - 4 - 5 1988 61
BC157915.1 - 3 - 4 1754 61
BC157957.1 - 3 - 4 1754 61
BC172121.1 - 3 - 4 1754 61

View Full Product Details