The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081315.1 | NP_001074784.1 | 9 - 10 | 3344 | 59 |
GenBank mRNA | AK129322.1 | - | 7 - 8 | 1599 | 59 |
AK147555.1 | - | 9 - 10 | 3343 | 59 | |
AK162638.1 | - | 2 - 3 | 192 | 59 | |
BC048580.1 | - | 2 - 3 | 221 | 59 | |
BC157915.1 | - | 8 - 9 | 3109 | 59 | |
BC157957.1 | - | 8 - 9 | 3109 | 59 |