Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lonrf3
Entrez Gene ID
74365
Gene Name
LON peptidase N-terminal domain and ring finger 3
Gene Aliases
4932412G04Rik, 5730439E01Rik, A830039N02Rik, AU023707, Rnf127
Chromosome Location
Chr.X: 36328358 - 36366857 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028894.1 NP_083170.1 3 - 4 1229 77
GenBank mRNA AK016522.1 - 3 - 4 1229 77
AK147751.1 - 3 - 4 1233 77
AK167341.1 - 3 - 4 1287 77
AK177253.1 - 3 - 4 294 77
AK189559.1 - 3 - 4 293 77
BC120688.1 - 3 - 4 1131 77
BC120690.1 - 3 - 4 1131 77

View Full Product Details