Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk20
Entrez Gene ID
105278
Gene Name
cyclin-dependent kinase 20
Gene Aliases
4932702G04Rik, AU019450, AW558027, CDCH, Ccrk, PNQLARE, p42
Chromosome Location
Chr.13: 64432363 - 64439722 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053180.2 NP_444410.1 1 - 2 123 100
GenBank mRNA AY005133.1 - 1 - 2 134 100
BC031907.1 - 1 - 2 123 100

View Full Product Details