Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Luc7l
Entrez Gene ID
66978
Gene Name
Luc7 homolog (S. cerevisiae)-like
Gene Aliases
1810045C04Rik, 2410018D03Rik
Chromosome Location
Chr.17: 26245364 - 26285510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025881.3 NP_080157.1 6 - 7 809 81
NM_028190.3 NP_082466.2 6 - 7 809 81
NR_037905.1 7 - 8 880 81
NR_037906.1 6 - 7 1866 81
GenBank mRNA AK010539.1 - 6 - 7 761 81
BC019194.1 - 7 - 8 802 81
BC034135.1 - 5 - 6 975 81
BC055875.1 - 6 - 7 1792 81
BQ572015.1 - 4 - 5 461 81
CF951283.1 - 2 - 3 205 81

View Full Product Details