The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028632.2 | NP_082908.2 | 4 - 5 | 322 | 151 |
GenBank mRNA | AK003561.1 | - | 3 - 4 | 281 | 151 |
BC033045.1 | - | 3 - 4 | 300 | 151 | |
BC094619.1 | - | 4 - 5 | 309 | 151 |