Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgef6
Entrez Gene ID
73341
Gene Name
Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6
Gene Aliases
1600028C08Rik, 1700038J06Rik, 4930592P22Rik, alpha-PIX
Chromosome Location
Chr.X: 57231485 - 57338756 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152801.2 NP_690014.2 2 - 3 479 138
GenBank mRNA AF393831.1 - 2 - 3 479 138
AK040468.1 - 2 - 3 301 138

View Full Product Details