Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lig3
Entrez Gene ID
16882
Gene Name
ligase III, DNA, ATP-dependent
Gene Aliases
D11Wsu78e
Chromosome Location
Chr.11: 82781109 - 82804274 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291245.1 NP_001278174.1 14 - 15 2288 84
NM_001291246.1 NP_001278175.1 14 - 15 2285 84
NM_001291247.1 NP_001278176.1 14 - 15 2289 84
NM_010716.3 NP_034846.2 14 - 15 2292 84
GenBank mRNA AK031540.1 - 14 - 15 2229 84
AK143531.1 - 14 - 15 2158 84
AK150644.1 - 14 - 15 2197 84
AK156018.1 - 14 - 15 2207 84
AK156030.1 - 3 - 4 274 84
AK156825.1 - 3 - 4 274 84
BC049240.1 - 14 - 15 2195 84
BC083500.1 - 14 - 15 2191 84
U66057.1 - 14 - 15 2226 87
U66058.1 - 14 - 15 2226 87

View Full Product Details