The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076789.1 | NP_001070257.1 | 1 - 2 | 87 | 68 |
GenBank mRNA | AK008792.1 | - | 1 - 2 | 123 | 68 |
BC004707.1 | - | 1 - 2 | 87 | 68 |