The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159509.1 | NP_001152981.1 | 8 - 9 | 1252 | 91 |
NM_001159510.1 | NP_001152982.1 | 8 - 9 | 1203 | 91 | |
NM_011862.3 | NP_035992.1 | 8 - 9 | 1291 | 91 | |
GenBank mRNA | AF128535.1 | - | 8 - 9 | 1281 | 91 |
AK154785.1 | - | 8 - 9 | 1178 | 91 | |
AK170299.1 | - | 8 - 9 | 1250 | 91 | |
AK184671.1 | - | 2 - 3 | 173 | 91 | |
BC023502.1 | - | 9 - 10 | 1157 | 91 | |
CF168957.1 | - | 3 - 4 | 396 | 91 |