Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smg7
Entrez Gene ID
226517
Gene Name
Smg-7 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
9430023P16Rik, mKIAA0250
Chromosome Location
Chr.1: 152836995 - 152902686 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005507.2 NP_001005507.1 7 - 8 815 79
NM_001160256.1 NP_001153728.1 7 - 8 815 79
NM_001160257.1 NP_001153729.1 7 - 8 805 79
GenBank mRNA AK129101.1 - 7 - 8 805 79
BC082789.1 - 7 - 8 802 79
BC086651.1 - 7 - 8 815 79

View Full Product Details