Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Angel1
Entrez Gene ID
68737
Gene Name
angel homolog 1 (Drosophila)
Gene Aliases
1110030H02Rik, mKIAA0759
Chromosome Location
Chr.12: 86700502 - 86734187 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144524.2 NP_653107.2 6 - 7 1564 91
GenBank mRNA AK143549.1 - 6 - 7 1565 91
BC019148.1 - 6 - 7 1579 91

View Full Product Details