Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dhrsx
- Entrez Gene ID
-
236082
- Gene Name
- dehydrogenase/reductase (SDR family) X chromosome
- Gene Aliases
-
C80877, D4Ertd296e, Dhrsxy, Pscad
- Chromosome Location
-
Chr.4|NT_166291: 15800 - 36213 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK030649.1 |
- |
4 - 5 |
404 |
113 |
View Full Product Details