Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl15
Entrez Gene ID
218639
Gene Name
ADP-ribosylation factor-like 15
Gene Aliases
A430036I03, Arfrp2, C230032K13Rik
Chromosome Location
Chr.13: 113794506 - 114157462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172595.4 NP_766183.1 1 - 2 186 64
GenBank mRNA AK039965.1 - 1 - 2 150 64
AK048736.1 - 1 - 2 69 64
AK050979.1 - 1 - 2 101 64
AK052164.1 - 1 - 2 135 64
BC120664.1 - 1 - 2 72 64
BC120666.1 - 1 - 2 72 64
BC144993.1 - 1 - 2 72 64
CJ063085.1 - 1 - 2 186 64
CJ066332.1 - 1 - 2 170 64

View Full Product Details