Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chadl
Entrez Gene ID
214685
Gene Name
chondroadherin-like
Gene Aliases
AY100452, D930017K21Rik
Chromosome Location
Chr.15: 81686166 - 81697376 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK086273.1 - - 866 62

View Full Product Details