The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011945.2 | NP_036075.2 | 19 - 20 | 4353 | 62 |
GenBank mRNA | AF117340.1 | - | 19 - 20 | 4353 | 62 |
L13103.1 | - | 9 - 10 | 2362 | 62 | |
U23470.1 | - | 6 - 7 | 1954 | 62 |