Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nell2
Entrez Gene ID
54003
Gene Name
NEL-like 2
Gene Aliases
A330108N19Rik, R75516, mel91
Chromosome Location
Chr.15: 95219440 - 95528758 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289653.1 NP_001276582.1 8 - 9 813 71
GenBank mRNA AK035019.1 - 6 - 7 666 71
AK039790.1 - - 877 71
AK048913.1 - - 1084 71
BC051968.1 - 7 - 8 781 71

View Full Product Details