Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Copz1
Entrez Gene ID
56447
Gene Name
coatomer protein complex, subunit zeta 1
Gene Aliases
5930435A22Rik, AA407760, D4Ertd360e
Chromosome Location
Chr.15: 103272834 - 103299865 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019817.1 NP_062791.1 4 - 5 269 115
GenBank mRNA AB037370.1 - 4 - 5 269 115
AK003302.1 - 4 - 5 277 115
AK079205.1 - 3 - 4 860 115
AK083824.1 - 4 - 5 289 115
AK156758.1 - 4 - 5 288 115
AK167636.1 - 4 - 5 277 115
AK168382.1 - 4 - 5 288 115
AK168471.1 - 4 - 5 288 115
BC002246.1 - 4 - 5 275 115
BC023207.1 - 3 - 4 253 115
BC025041.1 - 4 - 5 275 115
BC039920.1 - 4 - 5 298 115
BC052891.1 - 4 - 5 280 115
BC058524.1 - 4 - 5 268 115
BC062203.1 - 4 - 5 285 115
BC085314.1 - 4 - 5 290 115

View Full Product Details