Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pi16
Entrez Gene ID
74116
Gene Name
peptidase inhibitor 16
Gene Aliases
1200009H11Rik, Cripi, PI-16
Chromosome Location
Chr.17: 29318796 - 29328903 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023734.3 NP_076223.3 4 - 5 931 59
GenBank mRNA AB046537.1 - - 580 59
AI587910.1 - 2 - 3 210 59
BC039124.1 - 4 - 5 528 59
BC106088.1 - 4 - 5 616 59

View Full Product Details